Skip to Content
Merck
All Photos(1)

Documents

EHU008571

Sigma-Aldrich

MISSION® esiRNA

targeting human SRP54

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTTGAGCAATGCCACCATTATCAATGAAGAGGTATTGAATGCTATGCTAAAAGAAGTCTGTACCGCTTTGTTGGAAGCAGATGTTAATATTAAACTAGTGAAGCAACTAAGAGAAAATGTTAAGTCTGCTATTGATCTTGAAGAGATGGCATCTGGTCTTAACAAAAGAAAAATGATTCAGCATGCTGTATTTAAAGAACTTGTGAAGCTTGTAGACCCTGGAGTTAAGGCATGGACACCCACTAAAGGAAAACAAAATGTGATTATGTTTGTTGGATTGCAAGGGAGTGGTAAAACAACAACATGTTCAAAGCTAGCATATTATTACCAGAGGAAAGGTTGGAAGACCTGTTTAATATGTGCAGACACATTCAGAGCAGGGGCTTTTGACCAACTAAAACAGAATGCTACCAAAGCAAGAATTCCATTTTATGGAAGCTATACAGAAATGGATCCTGTCATCATTGCTTCTGAAGGAGTAGAGAAATTTAAAAATGAAAATTTTGAAATTATTATTGTTGATACAAGTGGCCG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abhik K Banerjee et al.
Cell, 183(5), 1325-1339 (2020-10-21)
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) is a recently identified coronavirus that causes the respiratory disease known as coronavirus disease 2019 (COVID-19). Despite the urgent need, we still do not fully understand the molecular basis of SARS-CoV-2 pathogenesis. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service