Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 20 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | € 212,00 | |
| 50 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | € 382,00 |
About This Item
€ 212,00
description
Powered by Eupheria Biotech
Quality Level
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
AAGACGTCGCTCTGGTTTGTGGGGTCTCTGCTGCTGGTGTCCGTCCTCATAGTCACCGTCGGGCTGGCTGCCACCACCAGGACGGAGAATGTGACCGTTGGGGGCTACTACCCAGGGATCATTCTCGGCTTTGGATCTTTCTTAGGAATTATTGGCATCAACTTGGTGGAGAATAGAAGGCAAATGCTGGTGGCAGCGATCGTGTTTATCAGTTTTGGCGTGGTGGCCGCCTTCTGCTGCGCCATCGTGGACGGCGTATTTGCAGCACAGCACATTGAACCGAGGCCCCTCACCACGGGAAGATGCCAGTTTTACTCCAGTGGGGTGGGGTACTTGTACGATGTCTACCAGACAGAGGTCACCTGTCACTCCCTGGACGGCAAGTGCCAGCTGAAGGTGAGAAGCAACACCTGTTACTGCTGTGACCTCTATGCCTG
Ensembl | human accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
human ... TMEM255B(348013), FAM70B(348013)
1 of 1
This Item | |||
|---|---|---|---|
| esiRNA cDNA target sequence AAGACGTCGCTCTGGTTTGTGGGGTCTCTGCTGCTGGTGTCCGTCCTCATAGTCACCGTCGGGCTGGCTGCCACCACCAGGACGGAGAATGTGACCGTTGGGGGCTACTACCCAGGGATCATTCTCGGCTTTGGATCTTTCTTAGGAATTATTGGCATCAACTTGGTGGAGAATAGAAGGCAAATGCTGGTGGCAGCGATCGTGTTTATCAGTTTTGGCGTGGTGGCCGCCTTCTGCTGCGCCATCGTGGACGGCGTATTTGCAGCACAGCACATTGAACCGAGGCCCCTCACCACGGGAAGATGCCAGTTTTACTCCAGTGGGGTGGGGTACTTGTACGATGTCTACCAGACAGAGGTCACCTGTCACTCCCTGGACGGCAAGTGCCAGCTGAAGGTGAGAAGCAACACCTGTTACTGCTGTGACCTCTATGCCTG | esiRNA cDNA target sequence CCAGGACCCAGAATGTGACTGTAGGAGGTTATTACCCCGGAGTTATTCTCGGCTTTGGATCGTTCCTTGGAATCATTGGATCAAACCTTATTGAGAACAAAAGGCAGATGCTGGTGGCTTCTATCGTGTTTATCAGCTTTGGTGTGATTGCGGCTTTTTGTTGTGCCATAGTTGACGGGGTCTTTGCTGCCAGACACATTGATCTGAAACCACTCTACGCTAACCGGTGCCATTATGTTCCCAAGACATCACAGAAGGAAGCTGAGGAGGTCATAAGTTCCTCAACCAAAAATTCTCCTTCCACGAGGGTTATGAGGAACCTTACCCAGGCAGCTAGAGAGGTTAACTGCCCTCACCTCAGCCGTGAATTCTGCACACCTCGCATCCGGGGCAACACCTGCTTCTGCTGTGACCTCTACAACTGTGGCAACCGGGTGGAGATCACTGGTGGGTACTACGAATACATCGATGTCAGCAGTTGCCAAGA | esiRNA cDNA target sequence CTGCTCCTCAGTGGCTTTTCGGCCTCAGAGGCCTCAGAGCCTGGTGTCATGTGGACTGGATATGCAGGTGATGCTGTGGAGTCTTCAAAAAGCCCGACCACTCTGGATTACAAATTTACAGGAGGATGAAACAGAAGAAATGGAAGGCCCACAGTCACCTGGTCAGCTCTTAAACCCTGCCCTAGCCCATTCTATCTCTGTGGCTTCGTGTGGTAATATTTTTAGTTGTGGTGCAGAAGATGGTAAGGTTCGAATCTTTCGGGTGATGGGAGTTAAGTGTGAACAGGAACTGGGATTTAAGGGCCACACTTCAGGGGTATCCCAGGTCTGCTTTCTCCCAGAATCCTATTTGCTGCTTACTGGAGGGAATGATGGGAAGATCACGTTGTGGGATGCAAACAGTG | esiRNA cDNA target sequence GACGACACTGCACTGAATGGGATCAGGCTGCACTGCGCGCGCGGGAACGTCCTAGGCAATACGCACGTGGTAGAGTCCCAGTCTGGAAGCTGGGGCGAATGGAGTGAGCCGCTGTGGTGTCGCGGCGGCGCCTACCTAGTGGCTTTCTCGCTTCGCGTGGAGGCACCCACGACCCTCGGTGACAACACAGCAGCGAACAACGTGCGCTTCCGCTGTTCAGACGGCGAGGAACTGCAGGGGCCTGGGCTGAGCTGGGGAGACTTTGGAGACTGGAGTGACCATTGCCCCAAGGGCGCGTGCGGCCTGCAGACCAAGATCCAGGGACCTAGAGGCCTCGGCGATGACACTGCGCTGAACGACGCGCGCTTATTCTGCTGCCGCAGTTGAACGGCGCCGCCGCCGCCGCTCTCTCCCGGGCCAGGAGGCTAGTCCCACCTC |
| Gene Information human ... TMEM255B(348013), FAM70B(348013) | Gene Information human ... TMEM255A(55026), FAM70A(55026) | Gene Information human ... WDR53(348793) | Gene Information human ... VMO1(284013) |
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| Ensembl | human accession no. | Ensembl | human accession no. | Ensembl | human accession no. | Ensembl | human accession no. |
| NCBI accession no. | NCBI accession no. | NCBI accession no. | NCBI accession no. |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
Storage Class
12 - Non Combustible Liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Global Trade Item Number
| SKU | GTIN |
|---|---|
| EHU006291-50UG | 04061828453030 |
| EHU006291-20UG | 04061828677153 |
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service