Skip to Content
Merck
All Photos(1)

Key Documents

EHU003471

Sigma-Aldrich

MISSION® esiRNA

targeting human SRF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACGACCTTCAGCAAGAGGAAGACGGGCATCATGAAGAAGGCCTATGAGCTGTCCACGCTGACAGGGACACAGGTGCTGTTGCTGGTGGCCAGTGAGACAGGCCATGTGTATACCTTTGCCACCCGAAAACTGCAGCCCATGATCACCAGTGAGACCGGCAAGGCACTGATTCAGACCTGCCTCAACTCGCCAGACTCTCCACCCCGTTCAGACCCCACAACAGACCAGAGAATGAGTGCCACTGGCTTTGAAGAGACAGATCTCACCTACCAGGTGTCGGAGTCTGACAGCAGTGGGGAGACCAAGGACACACTGAAGCCGGCGTTCACAGTCACCAACCTGCCGGGTACAACCTCCACCATCCAAACAGCACCTAGCACCTCTACCACCATGCAAGTCAGCAGCGGCCCCTCCTTTCCCATCACCAACTACCTGGCACC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xi He et al.
Oncology letters, 5(3), 819-824 (2013-02-22)
Recent studies indicate that serum response factor (SRF) is highly expressed in tumors such as hepatocellular, thyroid, esophageal and lung carcinoma. However, the expression and roles of SRF in esophageal squamous cell carcinoma (ESCC) are unclear. In this study, immunohistochemistry
Qi Li et al.
PloS one, 8(9), e75470-e75470 (2013-09-24)
Transcriptional regulation is essential for any gene expression including microRNA expression. MiR-1-1 and miR-133a-2 are essential microRNAs (miRs) involved in cardiac and skeletal muscle development and diseases. Early studies reveal two regulatory enhancers, an upstream and an intragenic, that direct
Carolina Leimgruber et al.
Journal of cellular physiology, 232(10), 2806-2817 (2016-11-20)
Prostatic smooth muscle cells (pSMCs) differentiation is a key factor for prostatic homeostasis, with androgens exerting multiple effects on these cells. Here, we demonstrated that the myodifferentiator complex Srf/Myocd is up-regulated by testosterone in a dose-dependent manner in primary cultures
Min-Seok Kim et al.
Scientific reports, 9(1), 4631-4631 (2019-03-16)
Methylmercury is an environmental pollutant that causes specific and serious damage to the central nervous system. We have previously shown that C-C motif chemokine ligand 4 (CCL4) protects cultured neural cells from methylmercury toxicity and expression of CCL4 is specifically
Allen Sam Titus et al.
American journal of physiology. Heart and circulatory physiology, 318(6), H1538-H1558 (2020-05-16)
Relative resistance to apoptosis and the ability to proliferate and produce a collagen-rich scar determine the critical role of cardiac fibroblasts in wound healing and tissue remodeling following myocardial injury. Identification of cardiac fibroblast-specific factors and mechanisms underlying these aspects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service