Skip to Content
Merck

Skip To

EC111

GenElute-E Tissue Stabilizer

greener alternative

Greener Alternative

Buffer for stabilization of samples for DNA or RNA purification. 100 or 500ML

Synonym(s):

Tissue Stabilizer

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.52
UNSPSC Code:
41105600


feature

Intended use (For stabilization of tissue samples prior to DNA or RNA purification)

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

room temp

General description

For storage of human or animal tissue samples and protection of DNA and RNA against degradation during storage. GenElute-E Tissue Stabilizer provides non-toxic stabilization of DNA or RNA isolated from tissue. Not intended for use with bacteria or cultured cells.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is aligned with "Waste prevention" and "Designing safer chemicals". Click here for more information.

Features and Benefits

Non-toxic stabilization of DNA or RNA isolated from tissue

Preparation Note

Use within 12 months of receipt.

Other Notes

We believe that green chemistry will contribute to a better tomorrow. With a growing portfolio of greener alternatives, there are now more choices to reduce the ecological impact of your research while still delivering quality and efficacy so your results are not compromised. GenElute-E kits adhere to the principles of SMASH Packaging, our global initiative to improve the sustainability through the reduction of packaging materials, the increased use of sustainable alternatives, and improved recyclability. Packaging is FCS-marked for sustainable forestry practices and produced from greater than 70% recycled content.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EC222EC700EC600
description

Buffer for stabilization of samples for DNA or RNA purification. 100 or 500ML

description

2X Loading dye for RNA gel electrophoresis. 500µL for 100 samples.

description

Reagents and consumables for organic solvent clean-up from DNA. 10, 50 or 250 purifications.

description

Reagents and consumables for clean-up of pre-purified DNA. 10, 50 or 250 purifications.

storage temp.

room temp

storage temp.

2-8°C

storage temp.

room temp

storage temp.

room temp

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

greener alternative category

Aligned,

greener alternative category

Aligned,

greener alternative category

, Aligned

greener alternative category

Aligned,

feature

Intended use (For stabilization of tissue samples prior to DNA or RNA purification)

feature

Intended use (For stabilization of RNA samples for RNA gel electrophoresis)

feature

Compatible Application (Suitable for most common downstream applications, including genotyping, PCR, and NGS), Intended use (For the removal of organic solvent traces from DNA solutions), Typical/expected yield (Varies by sample. Please reference user guide for more information.)

feature

Compatible Application (Suitable for most common downstream applications, including genotyping, PCR, and NGS), Intended use (For depletion of impurities and partial fractions (<50 bp) from DNA solutions), Typical/expected yield (Varies by sample. Please reference user guide for more information.)

sustainability

Greener Alternative Product

sustainability

Greener Alternative Product

sustainability

Greener Alternative Product

sustainability

Greener Alternative Product


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

12125-02-9

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters