Skip to Content

E9033

pFLAG-Myc-CMV-22 Expression Vector

Shuttle vector for transient or stable intracellular expression of dual tagged N-terminal FLAG and C-terminal c-myc fusion proteins

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


tag

FLAG® tagged, c-Myc tagged (C-terminal)

grade

Molecular Biology

form

buffered aqueous solution

shipped in

dry ice

storage temp.

−20°C

General description

The pFLAG-Myc-CMV-22 Expression Vector is a 6.3 kb derivative of pCMV5 used to establish transient or stable intracellular expression of dual-tagged N-terminal FLAG® and C-terminal c-myc fusion proteins in mammalian cells. The vector encodes the FLAG epitope (Asp-Tyr-Lys-Xaa-Xaa-Asp) and a c-myc epitope (EQKLISEEDL) upstream and downstream of the multiple cloning sites, respectively.

pFLAG-Myc-CMV-22 Expression Vector is a shuttle vector for E. coli and mammalian cells. Efficiency of replication is optimal when using an SV40 T antigen expressing host.

The pFLAG-CMV-4-BAP Control Plasmid is a 7.7 kb derivative of pCMV5 used for transient intracellular expression of N-terminal Met-FLAG bacterial alkaline phosphatase fusion protein in mammalian cells.

Vector Maps and Sequences

Application

The pFLAG-Myc-CMV-20 Expression Vector is suitable for stable cytoplasmic expression of C-terminal c-myc N-terminal Met-FLAG® fusion proteins in mammalian cells.

Biochem/physiol Actions

The promoter-regulatory region of the human cytomegalovirus drives transcription of FLAG® and c-myc fusion constructs. The aminoglycoside phosphotransferase II gene (Neo-r) confers resistance to aminoglycosides such as G418 allowing for selection of stable transfectants.

Other Notes

pFLAG-Myc-CMV-22 Expression Vector 20 μg (E5901) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.
pFLAG-CMV-4-BAP Control Plasmid 20 μg (C4722) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
pFLAG-CMV is a trademark of Sigma-Aldrich Co. LLC
pFLAG-Myc-CMV is a trademark of Sigma-Aldrich Co. LLC


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library