Skip to Content
Merck
All Photos(1)

Key Documents

E8651

Sigma-Aldrich

Anti-E2F3 antibody ,Mouse monoclonal

clone PG30, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing

Select a Size

0.2 ML
€685.00

€685.00


Estimated to ship on15 May 2025



Select a Size

Change View
0.2 ML
€685.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€685.00


Estimated to ship on15 May 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

PG30, monoclonal

form

buffered aqueous solution

mol wt

antigen 57 kDa

species reactivity

human

concentration

~2 mg/mL

technique(s)

immunoprecipitation (IP): suitable using denatured E2F3
microarray: suitable
western blot: 4-8 μg/mL using transfected 293T cells expressing E2F3

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... E2F3(1871)

General description

Monoclonal Anti-E2F3 (mouse IgG2a isotype) is derived from the PG30 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a recombinant human E2F Transcription Factor 3 (E2F3) protein. Transcription factor E2F3 is a DNA-binding protein of ~57 kDa encoded by the E2F3 gene in humans. It belongs to E2F transcription factor family of proteins that bind to the sequence TTTCGCGC.

Immunogen

recombinant human E2F3.

Application

Monoclonal Anti-E2F3 antibody produced in mouse has been used for the following applications:
  • immunoprecipitation using denatured E2F3
  • western blotting at a concentration of 4-8μg/mL using transfected 293T cells expressing E2F3
  • electrophoretic mobility shift assay (EMSA).

Biochem/physiol Actions

Transcription factor E2F3 is involved in the regulation of cell cycle. E2F3 helps in the regulation of the expression of a number of genes that are important in cell proliferation. It is a oncogene with strong proliferative potential and is involved in many processes. E2F3 plays an important role in the development of several types of cancer. Overexpression of E2F3 may be associated with hepatocellular carcinoma (HCC). It plays an important role in controlling cell cycle progression and proliferation. E2F3 is frequently expressed in pediatric Wilms′ tumors and may be associated with the same.

Physical form

Solution in phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.

Preparation Note

Purified from culture supernatant of hybridoma cells grown in a bioreactor.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cloning and characterization of human DP2, a novel dimerization partner of E2F.
Zhang, et al.
Oncogene, 10(11), 2085-2093 (1995)
Qun An et al.
Journal of pediatric surgery, 48(11), 2187-2193 (2013-11-12)
The transcription factor E2F3 plays an important role in controlling cell cycle progression and proliferation, and is overexpressed in various human cancers. The present study was undertaken to examine the expression of E2F3 and investigate its relevance in clinical and
Xiaoyun Zeng et al.
Oncology reports, 31(3), 1139-1146 (2014-01-10)
E2F transcription factor 3 (E2F3), a member of the E2F transcription factor family and a member of the genes involved in the regulation of cell cycle, is an oncogene with strong proliferative potential. E2F3 is involved in many processes and
Qiang Wang et al.
Immunity, inflammation and disease, 11(8), e987-e987 (2023-08-30)
E2F transcription factors are well-recognized oncogenic molecules, and their correlation with immune cell infiltration has recently been reported. This work studies the impacts and mechanism of E2F transcription factor 3 (E2F3) in the growth and tumor microenvironment (TME) of nasopharyngeal
J A Lees et al.
Molecular and cellular biology, 13(12), 7813-7825 (1993-12-01)
E2F is a transcription factor that helps regulate the expression of a number of genes that are important in cell proliferation. Recently, several laboratories have isolated a cDNA clone that encodes an E2F-like protein, known as E2F-1. Subsequent characterization of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service