Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
form
solid
Quality Segment
shipped in
ambient
storage temp.
room temp
SMILES string
[H][C@]12CC[C@@]3(C)[C@@]([H])(CC[C@@]3(O)C#C)[C@]1([H])CCc4cc(OC5CCCC5)ccc24
InChI
1S/C25H32O2/c1-3-25(26)15-13-23-22-10-8-17-16-19(27-18-6-4-5-7-18)9-11-20(17)21(22)12-14-24(23,25)2/h1,9,11,16,18,21-23,26H,4-8,10,12-15H2,2H3/t21-,22-,23+,24+,25+/m1/s1
InChI key
PWZUUYSISTUNDW-VAFBSOEGSA-N
Gene Information
human ... ESR1(2099), ESR2(2100)
1 of 1
This Item | |||
|---|---|---|---|
| description - | description ≥98% | description VETRANAL®, analytical standard | description ≥97.0% (HPLC) |
| form solid | form powder | form - | form powder |
| Quality Level 200 | Quality Level - | Quality Level 200 | Quality Level 200 |
| storage temp. room temp | storage temp. room temp | storage temp. - | storage temp. - |
| shipped in ambient | shipped in ambient | shipped in - | shipped in - |
| Gene Information human ... ESR1(2099), ESR2(2100) | Gene Information human ... ESR1(2099) | Gene Information - | Gene Information human ... AR(367) |
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Repr. 1B
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
PPE (personal protective equipment)
Eyeshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.





