Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
0.1 mg | Check Cart for Availability | € 478,00 |
About This Item
biological source
goat
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
species reactivity
mouse
technique(s)
flow cytometry: 0.25 μg using 106 cells, immunocytochemistry: 5-15 μg/mL, western blot: 1 μg/mL
UniProt accession no.
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
mouse ... Efnb2(13642)
General description
Immunogen
Biochem/physiol Actions
Physical form
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description affinity isolated antibody | description affinity isolated antibody, lyophilized powder | description affinity isolated antibody | description affinity isolated antibody, lyophilized powder |
| biological source goat | biological source goat | biological source rabbit | biological source goat |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| clone polyclonal | clone polyclonal | clone polyclonal | clone polyclonal |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



