Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.52
UNSPSC Code:
41106621
Quality Segment
storage temp.
−20°C
Application
For cytoplasmic expression
Legal Information
pFLAG-CMV is a trademark of Sigma-Aldrich Co. LLC
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.