Skip to Content
Merck

Skip To

E7280

Elafin

recombinant, expressed in Saccharomyces cerevisiae, powder, >90% (by MS, HPLC and SDS-PAGE),

Synonym(s):

Elastase-specific inhibitor, skin-derived antileukoprotease

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 765,00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
MDL number:

€ 765,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Elafin human, >90% (by MS, HPLC and SDS-PAGE), recombinant, expressed in Saccharomyces cerevisiae

Quality Segment

recombinant

expressed in Saccharomyces cerevisiae

assay

>90% (by MS, HPLC and SDS-PAGE)

form

powder

potency

1.7 x 10-10 M Ki (human leukocyte elastase), 4.2 x 10-10 M Ki (human proteinase 3)

mol wt

~6 kDa

UniProt accession no.

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

mode of action

enzyme | inhibits

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... PI3(5266)

General description

Chemical structure: peptide

Biochem/physiol Actions

Exhibits co-existent antimicrobial and antiproteolytic activities.1 Inhibits WAP-type four-disulfide core (WFDC) domain containing elastases. Antimicrobial spectrum: Gram-positive and Gram-negative bacteria.
Inhibitor of human leukocyte elastase and proteinase 3, originally isolated from the skin of patients with psoriasis. May protect against certain autoimmune tissue damage.
Inhibitor of human leukocyte elastase and proteinase 3, originally isolated from the skin of patients with psoriasis. May protect against certain autoimmune tissue damage. An elastase inhibitor which is an active fragment of elafin, as well as elafin itself, can also be found in bronchial carcinoma secretions.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
94261A6671M7152
description

>90% (by MS, HPLC and SDS-PAGE), recombinant, expressed in Saccharomyces cerevisiae

description

≥95% (HPLC)

description

-

description

≥97% (HPLC)

assay

>90% (by MS, HPLC and SDS-PAGE)

assay

≥95% (HPLC)

assay

≥98% (TLC)

assay

≥97% (HPLC)

Gene Information

human ... PI3(5266)

Gene Information

-

Gene Information

human ... ANPEP(290), DPP4(1803), ECE1(1889), LAP3(51056)

Gene Information

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder (Lyophilisate)

form

powder

form

-

shipped in

ambient

shipped in

-

shipped in

-

shipped in

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters