Skip to Content
Merck

E7275

Anti-Ephrin-A4 antibody produced in goat

affinity isolated antibody, lyophilized powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
51111800
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA (i), WB
Citations:
3
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

indirect ELISA: 0.5-1.0 μg/mL, western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EFNA4(1945)

General description

By ELISA, the antibody shows approximately 50% cross-reactivity with recombinant human ephrin-A5 and no cross-reactivity with recombinant mouse ephrin-A1, mouse ephrin-A2, human ephrin-A3, mouse ephrin-B1, mouse ephrin-B2, and mouse ephrin B3.

Immunogen

recombinant human ephrin-A4, extracellular domain, expressed in NSO cells.

Biochem/physiol Actions

Ephrin A4 binds to EPH receptor A4, a protein-tyrosine kinase family that belongs to the ephrin receptor subfamily. The ephrin-A and ephrin-B ligands and regulate developmental events, particularly those of the nervous system. EphrinA4 regulates the axonal growth, astrocyte reactivity and neuroinflammation in experimental autoimmune encephalomyelitis. High expression of EphA4 receptor correlates with tumor progression, invasion and metastasis of gastric cancer and is a suitable prognostic marker.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library