Skip to Content
Merck

E6515

Methionine Enkephalin-Arg-Gly-Leu

≥97% (HPLC)

Synonym(s):

MERGL, Tyr-Gly-Gly-Phe-Met-Arg-Gly-Leu

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C41H61N11O10S
CAS Number:
Molecular Weight:
900.06
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥97% (HPLC)

storage temp.

−20°C

Preparation Note

Originally extracted from bovine adrenal medulla


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library