Skip to Content
Merck

Skip To

E6155

pPolh-FLAG-2 Transfer Vector

Synonym(s):

ddddk tag, dykddddk tag

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 μg
Check Cart for Availability
€ 535,00

About This Item

NACRES:
NA.32
UNSPSC Code:
41106621

€ 535,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

shipped in

dry ice

storage temp.

−20°C

Application

Transfer vector for generation of recombinant Baculovirus and expression of C-terminal FLAG® fusions in insect cells.

Browse additional application references in our FLAG® Literature portal.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T6824F3040F2555
shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

baculovirus recombinant vector, N-terminal FLAG fusions, insect recombinant protein expression

baculovirus recombinant vector, N-terminal FLAG fusions, insect recombinant protein expression

-

-


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters