Skip to Content
Merck
All Photos(1)

Key Documents

E3750

Sigma-Aldrich

17-Epiestriol

Synonym(s):

1,3,5(10)-Estratriene-3,16α,17α-triol, 16α-Hydroxy-17α-estradiol, 3,16α,17α-Trihydroxy-1,3,5(10)-estratriene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24O3
CAS Number:
Molecular Weight:
288.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
form:
powder
Assay:
≥98.00% (TLC)

sterility

non-sterile

Quality Level

Assay

≥98.00% (TLC)

form

powder

technique(s)

inhibition assay: suitable

color

off-white to yellow

solubility

chloroform: methanol (1:1): 9.80-10.20 mg/mL, clear, colorless to faintly yellow

shipped in

ambient

storage temp.

room temp

SMILES string

CC12CCC3C(CCc4cc(O)ccc34)C1C[C@@H](O)[C@H]2O

InChI

1S/C18H24O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-17,19-21H,2,4,6-7,9H2,1H3/t13?,14?,15?,16-,17-,18?/m1/s1

InChI key

PROQIPRRNZUXQM-CKMBUZLOSA-N

Biochem/physiol Actions

17-epiestriol is an estradiol metabolite and a selective estrogen receptor (ER) beta agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carien van der Berg et al.
Analytical biochemistry, 590, 113531-113531 (2019-12-06)
An imbalance in the estrogen metabolism has been associated with an increased risk of breast cancer development. Evaluation of the estrogen biotransformation capacity requires monitoring of various estrogen metabolites. Up to now, only some estrogen metabolites could be measured in
Marcelo O Dietrich et al.
Trends in neurosciences, 36(2), 65-73 (2013-01-16)
The past 20 years witnessed an enormous leap in understanding of the central regulation of whole-body energy metabolism. Genetic tools have enabled identification of the region-specific expression of peripheral metabolic hormone receptors and have identified neuronal circuits that mediate the
Mitsuhiro Ueda et al.
Enzyme and microbial technology, 51(6-7), 402-407 (2012-10-09)
A protein with strong removal activity against the natural estrogen estriol was purified from a culture supernatant of Pleurotus eryngii var. tuoliensis C.J. Mou. The protein was characterized as a laccase and had a molecular mass of 60kDa on SDS-PAGE.
Swastika Surujlal-Naicker et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 48(1), 37-47 (2012-10-04)
Wastewater effluents have been documented as major contributors of hormone endocrine disrupting compounds (EDCs) in to the aquatic ecosystem. The need for rapid, simple and cost effective methods to detect these EDCs has increased. The use of Radio-immunoassays (RIA) were
Maya Frank et al.
Prenatal diagnosis, 33(2), 191-195 (2013-01-08)
To compare the profile of mid gestation triple test serum markers between a cohort of women with history of pregnancy complications with hereditary versus acquired thrombophilia. All were treated with low molecular weight heparin (LMWH) prior to 12 weeks' gestation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service