Skip to Content
Merck
All Photos(1)

Key Documents

E3256

Sigma-Aldrich

Edrophonium chloride

Synonym(s):

Ethyl(m-hydroxyphenyl)dimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16ClNO
CAS Number:
Molecular Weight:
201.69
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

[Cl-].CC[N+](C)(C)c1cccc(O)c1

InChI

1S/C10H15NO.ClH/c1-4-11(2,3)9-6-5-7-10(12)8-9;/h5-8H,4H2,1-3H3;1H

InChI key

BXKDSDJJOVIHMX-UHFFFAOYSA-N

Gene Information

human ... ACHE(43)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Acetycholinesterase inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leonardo Pisani et al.
ChemMedChem, 5(9), 1616-1630 (2010-08-03)
A large series of substituted coumarins linked through an appropriate spacer to 3-hydroxy-N,N-dimethylanilino or 3-hydroxy-N,N,N-trialkylbenzaminium moieties were synthesized and evaluated as acetylcholinesterase (AChE) and butyrylcholinesterase (BChE) inhibitors. The highest AChE inhibitory potency in the 3-hydroxy-N,N-dimethylanilino series was observed with a
Ulrike Schara et al.
Neuropediatrics, 43(4), 184-193 (2012-08-23)
Congenital myasthenic syndromes (CMS) are rare genetically and clinically heterogeneous disorders characterized by an impaired neuromuscular transmission. Exact prevalence data are not available, approximately 2000 to 3000 patients worldwide have been diagnosed on a molecular level; mutations in 14 different
Hariprasad Korsapati et al.
Gastroenterology, 135(3), 796-802 (2008-08-05)
Patients with high-amplitude esophageal contractions (nutcracker esophagus [NCE]) show asynchrony of circular muscle (CM) and longitudinal muscle (LM) contraction during peristalsis. The goal of our study was to determine if this asynchrony is related to an increase in the cholinergic
Manuel Martin-Flores et al.
Journal of the American Animal Hospital Association, 47(4), 294-298 (2011-06-16)
A case of prolonged muscle relaxation after vecuronium in an anesthetized dog is presented. After using peripheral nerve stimulation to confirm partial recovery of neuromuscular transmission, administration of 0.5 mg/kg IV of intravenous edrophonium failed to complete the reversal process.
Case report: A young man with progressive weakness, double vision and breathlessness.
Laura A Benjamin et al.
Malawi medical journal : the journal of Medical Association of Malawi, 23(3), 94-95 (2011-09-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service