E2126
Ethylenediamine dihydrochloride
Synonym(s):
1,2-Diaminoethane dihydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
NH2CH2CH2NH2 · 2HCl
CAS Number:
Molecular Weight:
133.02
Beilstein:
3665235
EC Number:
MDL number:
UNSPSC Code:
12352103
Recommended Products
mp
>300 °C (lit.)
SMILES string
Cl[H].Cl[H].NCCN
InChI
1S/C2H8N2.2ClH/c3-1-2-4;;/h1-4H2;2*1H
InChI key
OHHBFEVZJLBKEH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service