Sign In to View Organizational & Contract Pricing.
About This Item
EC Number:
209-842-5
UNSPSC Code:
12352200
density
1.36 g/mL at 25 °C (lit.)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.