Skip to Content
Merck
All Photos(1)

Key Documents

E0137

Sigma-Aldrich

Vanillic acid diethylamide

Synonym(s):

Ethamivan, N,N-Diethyl-4-hydroxy-3-methoxybenzamide, N,N-Diethylvanillamide

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H17NO3
CAS Number:
Molecular Weight:
223.27
Beilstein:
2694478
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

CCN(CC)C(=O)c1ccc(O)c(OC)c1

InChI

1S/C12H17NO3/c1-4-13(5-2)12(15)9-6-7-10(14)11(8-9)16-3/h6-8,14H,4-5H2,1-3H3

InChI key

BQJODPIMMWWMFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

CNS and respiratory stimulant.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

VANILLIC DIETHYLAMIDE (ETHAMIVAN) AND THE HYPOVENTILATION SYNDROME IN THORACIC SURGERY.
J P DECHENE
Canadian Anaesthetists' Society journal, 12, 173-178 (1965-03-01)
[EMIVAN, A USEFUL DRUG IN THE RECOVERY ROOM].
R R NOVOA
Revista espanola de anestesiologia, 11, 30-35 (1964-01-01)
A DOUBLE BLIND COMPARISON OF DOXAPRAM, ETHAMIVAN AND METHYLPHENIDATE.
B WOLFSON et al.
The American journal of the medical sciences, 249, 391-398 (1965-04-01)
Christopher W Murray et al.
Journal of medicinal chemistry, 53(16), 5942-5955 (2010-08-20)
Inhibitors of the chaperone Hsp90 are potentially useful as chemotherapeutic agents in cancer. This paper describes an application of fragment screening to Hsp90 using a combination of NMR and high throughput X-ray crystallography. The screening identified an aminopyrimidine with affinity
L S Rosenthal et al.
Biochemical pharmacology, 38(4), 581-588 (1989-02-15)
Carbon-13 nuclear magnetic resonance (NMR) techniques were employed to examine the effects of solvent environment on rotational barriers in two drugs known to cause widespread stimulation in the mammalian central nervous system: ethamivan and nikethamide. Total NMR bandshape analysis was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service