E001
Ethyl β-carboline-3-carboxylate
solid
Synonym(s):
β-CCE, Ethyl 9H-pyrido[3,4-b]indole-3-carboxylate
About This Item
form
solid
mp
228-230 °C (lit.)
solubility
ethanol: 3.5 mg/mL (warm)
DMSO: soluble
SMILES string
CCOC(=O)c1cc2c(cn1)[nH]c3ccccc23
InChI
1S/C14H12N2O2/c1-2-18-14(17)12-7-10-9-5-3-4-6-11(9)16-13(10)8-15-12/h3-8,16H,2H2,1H3
InChI key
KOVRZNUMIKACTB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
replaced by
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service