Key Documents
DWP96500Y4-EP
Eppendorf® Deepwell Plate 96/500μL
PCR Clean, volume 500 μL, yellow border with clear wells, pkg of 40 plates (5 bags x 8 plates)
About This Item
UNSPSC Code:
41122107
size:
96 well
sterility:
non-sterile
feature:
PCR Clean
yellow border with clear wells
Pricing and availability is not currently available.
sterility
non-sterile
feature
PCR Clean
yellow border with clear wells
packaging
pkg of 40 plates (5 bags x 8 plates)
manufacturer/tradename
Eppendorf® 951031828
availability
available only in North America
technique(s)
PCR: suitable
size
96 well
volume
500 μL
wells
96
Related Categories
General description
Eppendorf® Deepwell Plates 96 and 384 are high-performance plates for all manual and automated applications – from sample storage at -86 ºC to DNA denaturation at 100 ºC. Manufactured from pure polypropylene (PP), these plates offer innovative features: More dependability. More efficiency. More stability. The plates are available with a white, yellow, green, or blue border. For easy identification of wells, a high-contrast alphanumeric label is laser-engraved in the border. This OptiTrack® matrix allows you to find your sample up to 30 % more quickly while reducing pipetting errors. With all of these sophisticated features, these plates set new standards in your laboratory.
Features and Benefits
  • OptiTrack® matrix: up to 30 % faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling
  • RecoverMax® well design: Optimized well geometry for maximum sample recovery and excellent mixing properties
  • Minimal remaining volume and high uniformity from well to well
  • Raised well rims and smooth surface for reliable closing, even with heat sealing
  • Easy and reliable stacking
  • g-Safe: exceptional centrifugation stability up to 6,000 × g for faster protocols and improved sample quality
  • High-quality polypropylene guarantees high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without slip agents, plasticizers, and biocides – Materials which could have a negative effect on bioassays
  • Autoclavable (121?°C, 20?min)
  • Available with barcode. Additional details available at: www.eppendorf.com/barcode
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
RecoverMax is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)