Skip to Content
Merck
All Photos(4)

Key Documents

DUO92104

Sigma-Aldrich

Duolink® In Situ Orange Starter Kit Mouse/Goat

Synonym(s):

in situ Proximity Ligation Assay reagent, Protein Protein Interaction Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€1,310.00

€1,310.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€1,310.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

€1,310.00


Check Cart for Availability

Request a Bulk Order

product line

Duolink®

Quality Level

technique(s)

proximity ligation assay: suitable

fluorescence

λex 554 nm; λem 576 nm (orange) (Cyanine 3; Zeiss Filter set 20)

suitability

suitable for fluorescence

Application

Duolink®proximity ligation assay(PLA®) allows for endogenous detection of protein interactions, post translational modifications, and protein expression levels at the single molecule level in fixed cells and tissue samples.

Follow the Duolink® In Situ Fluorescence Protocol to use this product. A set of short instructionsis also available.

Visit our Duolink® PLA Resource Center for information on how to run a Duolink® experiment, applications, troubleshooting, and more.

To perform a complete Duolink® PLA in situ experiment you will need two primary antibodies (PLA, IHC, ICC or IF validated) that recognize two target epitopes. This starter kit supplies all other necessary reagents for 30 Duolink® PLA reactions, which include a pair of PLA probes (Anti-Mouse PLUS and Anti-Goat MINUS), orange detection reagents, wash buffers, and mounting medium.Note that the primary antibodies must come from the same species as the Duolink® PLA probes. Analysis is carried out using standard immunofluorescence assay equipment.
Specificity
The Duolink® In Situ Orange Starter Kit Mouse/Goat requires one primary antibody from mouse and one primary antibody from goat. Orange fluorescence detection reagents are often used with Cyanine 3 filter.

Application Note
Two primary antibodies raised in different species are needed. Test your primary antibodies (IgG-class, mono- or polyclonal) in a standard immunofluorescence (IF), immunohistochemistry (IHC) or immunocytochemistry (ICC) assay to determine the optimal fixation, blocking, and titer conditions. Duolink® PLA in situ reagents are suitable for use on fixed cells, cytospin cells, cells grown on slide, formalin-fixed, paraffin embedded (FFPE), or tissue (fresh or frozen). No minimum number of cells is required.

Let us do the work for you, learn more about our Custom Service Program to accelerate your Duolink® projects

View full Duolink® product list

Features and Benefits

  • No overexpression or genetic manipulation required
  • High specificity (fewer false positives)
  • Single molecule sensitivity due to rolling circle amplification
  • Relative quantification possible
  • No special equipment needed
  • Quicker and simpler than FRET
  • Increased accuracy compared to co-IP
  • Publication-ready results

Legal Information

Duolink is a registered trademark of Merck KGaA, Darmstadt, Germany
PLA is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Also Available Separately

Product No.
Description
SDS

  • DUO92006Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)SDS

  • DUO92001Duolink® In Situ PLA® Probe Anti-Mouse PLUSSDS

  • DUO92007Duolink® In Situ Detection Reagents OrangeSDS

  • DUO82049Duolink® In Situ Wash Buffers, FluorescenceSDS

  • DUO82040Duolink® In Situ Mounting Medium with DAPISDS

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Met. Corr. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gunisha Sagar et al.
Gut, 65(7), 1165-1174 (2015-06-11)
New-onset diabetes and concomitant weight loss occurring several months before the clinical presentation of pancreatic cancer (PC) appear to be paraneoplastic phenomena caused by tumour-secreted products. Our recent findings have shown exosomal adrenomedullin (AM) is important in development of diabetes
Linbo Wang et al.
Journal of cancer research and clinical oncology, 140(12), 1997-2008 (2014-07-10)
Our previous work identified leucine zipper transcription factor-like 1 (LZTFL1) as a novel tumor suppressor gene, with its expression correlated with survival outcome in gastric cancer (GC) patients. This study focuses on the role of LZTFL1 in GC aggression and
Shaida Ouladan et al.
International journal of oncology, 46(6), 2595-2605 (2015-04-23)
Solitary fibrous tumors (SFTs) are rare mesenchymal neoplasms, displaying variable morphological and clinicopathological features. Supportive immunohistochemical markers such as CD34, CD99, BCL2 and LSD1 are commonly applied in the differential diagnosis of SFTs, although none is sufficiently sensitive or specific
Dipti Pravin Lambade et al.
Journal of clinical and diagnostic research : JCDR, 9(2), ZC01-ZC05 (2015-04-11)
The purpose of this vitro study was to comparatively evaluate the adhesive bonding of dual cured resin luting agents with lithium disilicate ceramic material. Porcelain laminate veneers were prepared with lithium disilicate ceramic material i.e. IPS Empress II( E-Max Press).
Xiaodong Xu et al.
Zhonghua kou qiang yi xue za zhi = Zhonghua kouqiang yixue zazhi = Chinese journal of stomatology, 49(11), 677-681 (2015-01-28)
To evaluate and compare the physical and chemical properties of four different dental cements under long-term water storage. A glass-ionomer cement (A:Fuji I), a resin reinforced glass-ionomer (B: Fuji Plus), a self-adhesive resin cement (C:G-Cem), and an etch & rinse

Articles

Duolink® proximity ligation assay used to study neuron interactions furthering neuroscience research.

Things to consider for preparation, setup and execution of the Duolink® assay protocol

Support information including tips and tricks, frequently asked questions, and basic troubleshooting.

Protocols

This page details the Duolink® In Situ Short Protocol for fluorescence detection

This protocol describes how to perform immunofluorescent detection of proteins in cells and tissue.

Related Content

Applications to detect, quantify and visualize protein-protein interactions, post-translational modifications and low expression protein detection using proximity ligation assay

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service