Skip to Content
Merck

D9504

DNP-glycine

Synonym(s):

N-(2,4-Dinitrophenyl)glycine

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C8H7N3O6
CAS Number:
Molecular Weight:
241.16
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
214-114-5
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI key

INIGODASXCUILV-UHFFFAOYSA-N

InChI

1S/C8H7N3O6/c9-7(8(12)13)5-2-1-4(10(14)15)3-6(5)11(16)17/h1-3,7H,9H2,(H,12,13)

SMILES string

OC(CNC1=C([N+]([O-])=O)C=C([N+]([O-])=O)C=C1)=O

assay

≥98%

form

powder

technique(s)

particle immunofluorescence: suitable

color

light yellow to yellow

storage temp.

−20°C

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luisa A Ferreira et al.
Journal of biomolecular structure & dynamics, 35(1), 207-218 (2016-01-05)
Solvent properties of aqueous media (dipolarity/polarizability, hydrogen bond donor acidity, and hydrogen bond acceptor basicity) were measured in the coexisting phases of Dextran-PEG aqueous two-phase systems (ATPSs) containing .5 and 2.0 M urea. The differences between the electrostatic and hydrophobic properties

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service