Skip to Content
Merck
All Photos(1)

Key Documents

D7446

Sigma-Aldrich

Phenoxodiol

≥98% (HPLC)

Synonym(s):

3-(4-Hydroxyphenyl)-2H-1-benzopyran-7-ol, Dehydroequol, Haginin E, Idronoxil, NV 06

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O3
CAS Number:
Molecular Weight:
240.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

SMILES string

Oc1ccc(cc1)C2=Cc3ccc(O)cc3OC2

InChI

1S/C15H12O3/c16-13-4-1-10(2-5-13)12-7-11-3-6-14(17)8-15(11)18-9-12/h1-8,16-17H,9H2

InChI key

ZZUBHVMHNVYXRR-UHFFFAOYSA-N

Biochem/physiol Actions

Phenoxodiol is a Pan-cancer drug; causes apoptosis via both intrinsic and extrinsic pathways; targets plasma membrane electron transport (PMET).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan B Howes et al.
BMC clinical pharmacology, 11, 1-1 (2011-02-05)
Phenoxodiol is a novel isoflavone currently being studied in clinical trials for the treatment of cancer. This study reports the pharmacokinetics of phenoxodiol in patients with cancer. The pharmacokinetics of phenoxodiol was studied following a single intravenous (iv) bolus dose
Kate Porter et al.
Current cancer drug targets, 20(5), 341-354 (2020-01-04)
Idronoxil has been the subject of more than 50 peer-reviewed publications over the last two decades. This isoflavone is an intriguing regulator of multiple signal transduction pathways, capable of causing a range of biological effects, including cell cycle arrest, apoptosis
R A C McPherson et al.
British journal of cancer, 100(4), 649-655 (2009-02-12)
Phenoxodiol is a novel isoflav-3-ene, currently undergoing clinical trials, that has a broad in vitro activity against a number of human cancer cell lines. Phenoxodiol alone inhibited DU145 and PC3 in a dose- and time-dependent manner with IC(50) values of
Simon Mahoney et al.
Cancer cell international, 14(1), 110-110 (2014-11-18)
Prostate cancer is associated with a poor survival rate. The ability of cancer cells to evade apoptosis and exhibit limitless replication potential allows for progression of cancer from a benign to a metastatic phenotype. The aim of this study was
Harriet M Kluger et al.
Journal of translational medicine, 5, 6-6 (2007-01-30)
XIAP up-regulation is associated with chemotherapy resistance. Phenoxodiol causes XIAP degradation and chemotherapy sensitization in ovarian cancer. Here we assessed XIAP expression in melanomas, using tissue microarrays containing 436 melanomas and 336 nevi by a novel method of automated, quantitative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service