Skip to Content

D7191

Dibenzylfluorescein

Synonym(s):

DBF

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C34H24O5
Molecular Weight:
512.55
UNSPSC Code:
12171500
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99-102% (NMR)

Quality Segment

form

solid

shipped in

wet ice

storage temp.

−20°C

SMILES string

O=C1C=CC2=C(c3ccc(OCc4ccccc4)cc3OC2=C1)c5ccccc5C(=O)OCc6ccccc6

InChI

1S/C34H24O5/c35-25-15-17-29-31(19-25)39-32-20-26(37-21-23-9-3-1-4-10-23)16-18-30(32)33(29)27-13-7-8-14-28(27)34(36)38-22-24-11-5-2-6-12-24/h1-20H,21-22H2

InChI key

YZJGKSLPSGPFEV-UHFFFAOYSA-N

Application

DBF is dealkylated by cytochromes P450 (especially CYP2C8, CYP2C9, CYP2C19, and CYP3A) to form a fluorescein ester. Dibenzylfluorescein was reported to be suitable for in vitro assessment of the initial screening of CYP3A4-
mediated drug interactions. The fluorescent fluorescein product can be quantitated after the addition of base, which hydrolyzes the ester and increases the pH for optimal fluorescein fluorescence detection.
DBF is dealkylated by cytochromes P450 (especially CYP2C8, CYP2C9, CYP2C19, and CYP3A) to form a fluorescein ester. The fluorescent fluorescein product can be quantitated after the addition of base, which hydrolyzes the ester and increases the pH for optimal fluorescein fluorescence detection.

Other Notes

Dibenzylfluorescein is light sensitive and should be protected from direct sunlight or UV sources.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library