Skip to Content
Merck

Skip To

D6946

(R)-DRF053 hydrochloride hydrate

≥98% (HPLC)

Synonym(s):

2-[[9-(1-Methylethyl)-6-[[3-(2-pyridinyl)phenyl]amino]- 9H-purin-2-yl]amino]-1-butanol hydrochloride hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 418,00

About This Item

Empirical Formula (Hill Notation):
C23H27N7O · xHCl · yH2O
Molecular Weight:
417.51 (anhydrous free base basis)
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder

€ 418,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

yellow

solubility

DMSO: ≥30 mg/mL

storage temp.

2-8°C

SMILES string

O.Cl.CC[C@H](CO)Nc1nc(Nc2cccc(c2)-c3ccccn3)c4ncn(C(C)C)c4n1

InChI

1S/C23H27N7O.ClH.H2O/c1-4-17(13-31)27-23-28-21(20-22(29-23)30(14-25-20)15(2)3)26-18-9-7-8-16(12-18)19-10-5-6-11-24-19;;/h5-12,14-15,17,31H,4,13H2,1-3H3,(H2,26,27,28,29);1H;1H2/t17-;;/m1../s1

InChI key

ZOSNTFVWVJFXQO-ZEECNFPPSA-N

Biochem/physiol Actions

DRF053 is a more potent version of roscovitine.
DRF053 is a more potent version of roscovitine. It is 2 fold more potent against CDK5 (80 nM) and over 150 times more potent against CDK1 (14 nM).

Features and Benefits

This compound is featured on the CDKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
R2533SML0191P5234
form

powder

form

powder

form

powder

form

solid

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥97% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

solubility

DMSO: ≥30 mg/mL

solubility

DMSO: ≥20 mg/mL

solubility

H2O: ≥8 mg/mL

solubility

DMSO: soluble 37 mg/mL, H2O: insoluble

color

yellow

color

white to tan

color

white to tan

color

-


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters