Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C14H17N5O6
CAS Number:
Molecular Weight:
351.31
UNSPSC Code:
12352202
PubChem Substance ID:
EC Number:
249-929-5
MDL number:
assay
98-99% (TLC)
form
solid
storage temp.
−20°C
SMILES string
O=C(C)O[C@H]1[C@@H](OC(C)=O)[C@H](N2C3=NC=NC(N)=C3N=C2)O[C@@H]1CO
InChI
1S/C14H17N5O6/c1-6(21)23-10-8(3-20)25-14(11(10)24-7(2)22)19-5-18-9-12(15)16-4-17-13(9)19/h4-5,8,10-11,14,20H,3H2,1-2H3,(H2,15,16,17)/t8?,10-,11-,14-/m1/s1
InChI key
OUQFHKYWNKYTTQ-PQKOYNMZSA-N
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.