Skip to Content
Merck

Skip To

D6162

DKW/Juglans Basal Salt Mixture

powder, suitable for plant cell culture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 L
Check Cart for Availability
€ 17,50

About This Item

NACRES:
NA.72
UNSPSC Code:
12352207

€ 17,50


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

technique(s)

cell culture | plant: suitable

application(s)

agriculture

storage temp.

2-8°C

Application

DKW/Juglans (walnut) Basal Salt Mixture is composed of macro- and micronutrients. DKW/Juglans Basal Salt Mixture may be used as a chemstock to support the reconstitution of DKW/Juglans medium. DKW medium was originally developed for walnut micropropagation.

Media Formulation
DKW/Juglans Basal Salt Mixture has been used as a basal medium for the growth of leaf explants.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S6765C1416M2909
technique(s)

cell culture | plant: suitable

technique(s)

cell culture | plant: suitable

technique(s)

cell culture | plant: suitable

technique(s)

cell culture | plant: suitable

form

powder

form

fine powder

form

powder

form

powder

application(s)

agriculture

application(s)

agriculture

application(s)

agriculture

application(s)

agriculture

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Ox. Sol. 3

Storage Class

5.1B - Oxidizing hazardous materials

wgk

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

6484-52-2

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. May I request for the compositions of this DKW basal salt

    1 answer
    1. The composition of this product, DKW/Juglans Basal Salt Mixture, can be viewed at: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/361/201/d6162for.

      Helpful?

Reviews

No rating value

Active Filters