Skip to Content
Merck
All Photos(1)

Key Documents

D5321

Sigma-Aldrich

D36 hydrate

≥98% (HPLC)

Synonym(s):

5-(2-methoxyphenyl)-1-(2-methylpropyl)-1H-Indole-3-acetamide Hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24N2O2 · xH2O
CAS Number:
Molecular Weight:
336.43 (anhydrous basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: >10 mg/mL

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

O.COc1ccccc1-c2ccc3n(CC(C)C)cc(CC(N)=O)c3c2

InChI

1S/C21H24N2O2.H2O/c1-14(2)12-23-13-16(11-21(22)24)18-10-15(8-9-19(18)23)17-6-4-5-7-20(17)25-3;/h4-10,13-14H,11-12H2,1-3H3,(H2,22,24);1H2

InChI key

ZERKAKHVUQMJPW-UHFFFAOYSA-N

Biochem/physiol Actions

D36 is an androgen receptor (AR) allosteric antagonist with inverse agonist properties. D36 inhibits both wild-type and agonist-induced AR activity and antagonized AR in a hormone-refractory model.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service