Skip to Content
Merck
All Photos(1)

Key Documents

D4500

Supelco

Dehydroisoandrosterone 3-acetate

Synonym(s):

3-β-Acetoxy-5-androsten-17-one, Prasterone acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O3
CAS Number:
Molecular Weight:
330.46
EC Number:
MDL number:
UNSPSC Code:
41116107

drug control

Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

168-170 °C (lit.)

format

neat

SMILES string

CC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CCC4=O)[C@@H]3CC=C2C1

InChI

1S/C21H30O3/c1-13(22)24-15-8-10-20(2)14(12-15)4-5-16-17-6-7-19(23)21(17,3)11-9-18(16)20/h4,15-18H,5-12H2,1-3H3/t15-,16-,17-,18-,20-,21-/m0/s1

InChI key

NCMZQTLCXHGLOK-ZKHIMWLXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Leighton et al.
The Journal of nutrition, 117(7), 1287-1290 (1987-07-01)
The present study is a follow-up to the report that dehydroepiandrosterone (DHEA) acetate treatment in rats stimulated metabolic heat production and suppressed serum triglycerides, adiposity and weight gain without affecting food intake. Activities of peroxisomal fatty acyl-coenzyme (CoA) oxidase and
D C Cumming
Cleveland Clinic journal of medicine, 57(2), 161-166 (1990-03-01)
The effects of estrogenic steroid hormones in women are apparent, but the circulating androgen levels are much higher. Compared with serum estrone and estradiol levels, circulating testosterone levels are five to 10 times higher, androstenedione levels 30 times higher, dehydroepiandrosterone
H Shinohara et al.
Experientia, 36(11), 1329-1330 (1980-11-15)
Degeneration of ovarian oocytes occurred to a remarkable extent in rats with polycystic ovaries induced by dehydroepiandrosterone acetate (DHA-Ac) administration. The ratio of degeneration oocytes, compared with the total oocytes examined, finally exceeded 70%.
M A Graf et al.
Fertility and sterility, 59(4), 761-767 (1993-04-01)
To elucidate changes in gonadotropin secretion pattern in patients with hyperandrogenemic chronic anovulation of various origins. Hyperandrogenemic patients (n = 32), divided into subgroups according to certain clinical and biochemical criteria, and a control group (n = 9) of regularly
Y Shimomura et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 19(7), 295-299 (1987-07-01)
Adrenalectomy in young obese (ob/ob) and the diabetic (db/db) mouse slowed body weight gain. Treatment of adrenalectomized ob/ob mice with cortisone or deoxycorticosterone acetate (DOCA) significantly increased weight gain in a dose-related manner. Cortisone had no effect on weight gain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service