Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C15H12I2O4
CAS Number:
Molecular Weight:
510.06
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
storage temp.
2-8°C
SMILES string
OC(=O)CCc1cc(I)c(Oc2ccc(O)cc2)c(I)c1
Biochem/physiol Actions
DITPA, an analog of L-thyroxine, has inotropic but not chronotropic effects on heart muscle. It also stimulates myocardial angiogenesis after infarction by interaction with integrin αvཪ3 at the cell surface.
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class
13 - Non Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
PPE (personal protective equipment)
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
