D3909
cis-13,16,19-Docosatrienoic acid methyl ester
≥97.0% (capillary GC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C23H40O2
CAS Number:
Molecular Weight:
348.56
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
Assay
≥97.0% (capillary GC)
form
liquid
functional group
ester
shipped in
ambient
storage temp.
−20°C
SMILES string
CC\C=C/C\C=C/C\C=C/CCCCCCCCCCCC(=O)OC
InChI
1S/C23H40O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23(24)25-2/h4-5,7-8,10-11H,3,6,9,12-22H2,1-2H3/b5-4-,8-7-,11-10-
InChI key
RXNIYBHTXVTSAK-YSTUJMKBSA-N
Packaging
Sealed ampule
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service