Skip to Content
Merck
All Photos(3)

Key Documents

D3813

Sigma-Aldrich

Anti-DR4, N-Terminal antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Death Receptor 4, Anti-TRAIL-R1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen approximately 57 kDa

species reactivity

human

technique(s)

microarray: suitable
western blot: 0.5 μg/mL using human HeLa cell extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

General description

DR4, referred to as TRAIL Receptor-1, is a novel death domain containing receptor whose ligand has been identified as TRAIL or apoptosis-inducing ligand 2 (Apo2L). It is a member of the TNF superfamily of receptors that induce apoptosis. It is expressed in most human tissues including spleen, peripheral blood leukocytes, small intestine, and thymus.

Immunogen

synthetic peptide corresponding to amino acids 1-20 of the N-terminal of the mature human DR4.

Application

Anti-DR4 antibody produced in rabbit is suitable for microarray and immunoblotting at a working dilution of 1:500-1:1000 using total HeLa, K562, and Jurkat whole cell lysates. It was used as a primary antibody in the immunodetection of DR4 in cell lysate of human small cell lung carcinoma cells. It was used as a primary antibody in the immunoblot analysis of fetal and adenocarcinoma epithelial colon cells in a study.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

DR4 induces apoptosis in a variety of human tumor cell lines, but not in normal cells and activates NF-κB.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lenka Kočí et al.
Cytokine, 55(1), 34-39 (2011-04-13)
The resistance of transformed epithelial cells to a detachment-induced apoptosis (anoikis) can significantly affect their susceptibility to anticancer therapy. We showed that detachment of both fetal (FHC) and adenocarcinoma (HT-29) human colon epithelial cells resulted in the activation of the
P Schneider et al.
Immunity, 7(6), 831-836 (1998-01-16)
TRAIL induces apoptosis through two closely related receptors, TRAIL-R1 (DR4) and TRAIL-R2 (DR5). Here we show that TRAIL-R1 can associate with TRAIL-R2, suggesting that TRAIL may signal through heteroreceptor signaling complexes. Both TRAIL receptors bind the adaptor molecules FADD and
Alena Vaculova et al.
Molecular cancer, 9, 87-87 (2010-04-27)
TRAIL is considered as a promising anti-cancer agent, because of its ability to induce apoptosis in cancer but not in most normal cells. However, growing evidence exist that many cancer cells are resistant to its apoptotic effects. SCLC is a
S R Wiley et al.
Immunity, 3(6), 673-682 (1995-12-01)
A novel tumor necrosis factor (TNF) family member has been cloned and characterized. This protein, designated TNF-related apoptosis-inducing ligand (TRAIL), consists of 281 and 291 aa in the human and murine forms, respectively, which share 65% aa identity. TRAIL is
G Pan et al.
Science (New York, N.Y.), 276(5309), 111-113 (1997-04-04)
TRAIL (also known as Apo-2L) is a member of the tumor necrosis factor (TNF) ligand family that rapidly induces apoptosis in a variety of transformed cell lines. The human receptor for TRAIL was found to be an undescribed member of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service