Skip to Content
Merck
All Photos(1)

Key Documents

D2661

Sigma-Aldrich

cis-4,7,10,13,16,19-Docosahexaenoic acid ethyl ester

~99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H36O2
CAS Number:
Molecular Weight:
356.54
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~99%

storage temp.

−20°C

SMILES string

CCOC(=O)CC\C=C\C\C=C\C\C=C\C\C=C\C\C=C\C\C=C\CC

InChI

1S/C24H36O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24(25)26-4-2/h5-6,8-9,11-12,14-15,17-18,20-21H,3-4,7,10,13,16,19,22-23H2,1-2H3/b6-5+,9-8+,12-11+,15-14+,18-17+,21-20+

InChI key

ITNKVODZACVXDS-YATCGRJWSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule.

comparable product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J H Song et al.
Atherosclerosis, 155(1), 9-18 (2001-02-27)
The effect of dietary docosahexaenoic acid (DHA, 22:6n-3) oil with different lipid types on lipid peroxidation was studied in rats. Each group of male Sprague-Dawley rats was pair fed 15% (w/w) of either DHA-triglycerides (DHA-TG), DHA-ethyl esters (DHA-EE) or DHA-phospholipids
Alfonso Valenzuela et al.
Annals of nutrition & metabolism, 49(1), 49-53 (2005-03-01)
Docosahexaenoic acid (C22:6, DHA) is an omega-3 fatty acid required for the normal development of the mammalian nervous and visual system. DHA is provided by the mother during pregnancy and lactating period. Mother's DHA supplementation during pregnancy, and even before
M Martinez
Journal of molecular neuroscience : MN, 16(2-3), 309-316 (2001-08-02)
Patients with the Zellweger syndrome and its variants have very low levels of docosahexaenoic acid (DHA) in the brain, retina, and other tissues. Such a marked DHA deficiency could be related to the pathogenesis of peroxisomal disorders. Therefore, restoring the
L Poisson et al.
Journal of biotechnology, 91(1), 75-81 (2001-08-28)
In order to produce docosahexaenoic acid (DHA), a culture of the microalgal strain Isochrysis galbana was implemented. In Erlenmeyer flasks, a natural seawater medium, the Provasoli 1/3 medium, was compared to the classical Jones medium for DHA production. The Provasoli
K Umegaki et al.
Free radical research, 34(4), 427-435 (2001-05-01)
We compared the influence of docosahexaenoic acid (DHA) supplementation on oxidative DNA damage in bone marrow between young and aged rats. As a marker of oxidative DNA damage, 8-hydroxydeoxyguanosine (8-OHdG) in DNA was analyzed. Young (5-week-old) and aged (100-week-old) female

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service