Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C26H35NO4
CAS Number:
Molecular Weight:
425.56
UNSPSC Code:
41116107
EC Number:
238-325-7
MDL number:
drug control
USDEA Schedule II; regulated under CDSA - not available from Sigma-Aldrich Canada
InChI
1S/C26H35NO4.ClH/c1-23(2,29)18-13-24-8-9-26(18,30-3)22-25(24)10-11-27(14-15-4-5-15)19(24)12-16-6-7-17(28)21(31-22)20(16)25;/h6-7,15,18-19,22,28-29H,4-5,8-14H2,1-3H3;1H/t18-,19-,22-,24?,25+,26?;/m1./s1
InChI key
UNPUINKKJUSSDZ-NNGCTRGCSA-N
General description
Diprenorphine is a non-selective and an opiate receptor antagonist, that finds applications in veterinary medicine, in reversing the effects of opioid analgesics used for tranquilizing large animals.
Biochem/physiol Actions
Potent nonselective opiod antagonist.
signalword
Warning
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Veterinary Anesthesia and Analgesia: The Fifth Edition of Lumb and Jones (2015)
New Trends in Radiopharmaceutical Synthesis, Quality Assurance, and Regulatory Control (2013)
Zoo Animal and Wildlife Immobilization and Anesthesia (2014)
