Skip to Content
Merck

D181

D3 Dopamine Receptor from rat

recombinant, expressed in E. coli, membrane suspension

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352116
NACRES:
NA.32
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

recombinant

expressed in E. coli

form

membrane suspension

packaging

vial of 50 units

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

rat ... Drd3(29238)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
S2067I8657A6853
recombinant

expressed in E. coli

recombinant

-

recombinant

expressed in bacteria

recombinant

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

form

membrane suspension

form

-

form

lyophilized powder

form

membrane suspension

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

Gene Information

rat ... Drd3(29238)

Gene Information

rat ... Mrps9(301371)

Gene Information

rat ... Ifna_mapped(24480)

Gene Information

-

Physical form

Membrane suspension in 50 mM Tris-HCl, pH 7.4, 10% glycerol, 1% BSA.

Analysis Note

Recommended assay conditions are supplied on the product data sheet. This is a bioassayed product. Actual concentration and specific binding capacity are provided with each lot.

Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M L Bouthenet et al.
Brain research, 564(2), 203-219 (1991-11-15)
The messenger RNA (mRNA) of the recently characterized D3 dopamine receptor was visualized on rat brain sections using in situ hybridization with a 32P-labeled ribonucleic acid probe corresponding to a major part of the third cytoplasmic loop, a domain in
L Tang et al.
The Journal of pharmacology and experimental therapeutics, 268(1), 495-502 (1994-01-01)
In order to study the properties of the D2-like dopamine receptors, D2, D3 and D4 clones were transfected into mouse Ltk- fibroblasts, CCL1.3, and a neuronal mesencephalic cell line, MN9D. Most of the derived antagonist and agonist inhibition constants were
P Sokoloff et al.
Nature, 347(6289), 146-151 (1990-09-13)
A dopamine receptor has been characterized which differs in its pharmacology and signalling system from the D1 or D2 receptor and represents both an autoreceptor and a postsynaptic receptor. The D3 receptor is localized to limbic areas of the brain

Related Content

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service