Skip to Content
Merck

Skip To

D1393

4-(N-[2,4-Diamino-6-pteridinylmethyl]amino)benzoic acid sodium salt

≥95% (TLC)

Synonym(s):

4-Amino-4-deoxypteroic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 mg
Check Cart for Availability
€ 371,00
€ 278,25

About This Item

Linear Formula:
C14H12N7O2Na
CAS Number:
Molecular Weight:
333.28
NACRES:
NA.28
PubChem Substance ID:
UNSPSC Code:
12352106
MDL number:

€ 278,25

List Price€ 371,00Save 25%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

assay

≥95% (TLC)

form

powder

color

yellow to orange

SMILES string

[Na+].Nc1nc(N)c2nc(CNc3ccc(cc3)C([O-])=O)cnc2n1

InChI

1S/C14H13N7O2.Na/c15-11-10-12(21-14(16)20-11)18-6-9(19-10)5-17-8-3-1-7(2-4-8)13(22)23;/h1-4,6,17H,5H2,(H,22,23)(H4,15,16,18,20,21);/q;+1/p-1

InChI key

NDXULQWYAMJQGC-UHFFFAOYSA-M

Application

4-(N-[2,4-Diamino-6-pteridinylmethyl]amino)benzoic acid is an impurity found in methotrexate preparations. It may be used as a reference compound in methothrexate purity analysis.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
861553718181H9633
biological source

synthetic (organic)

biological source

-

biological source

-

biological source

-

assay

≥95% (TLC)

assay

95%

assay

≥95%

assay

≥95%, ≥98% (TLC)

form

powder

form

solid

form

solid

form

powder

color

yellow to orange

color

-

color

-

color

white to off-white

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters