Skip to Content
Merck
All Photos(1)

Key Documents

D0942

Sigma-Aldrich

Anti-DHFR, C-terminal antibody produced in rabbit

1.0-1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Dihydrofolate reductase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€490.00

€490.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€490.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.56

€490.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

concentration

1.0-1.5 mg/mL

technique(s)

immunoprecipitation (IP): 0.5-1.0 μg using 100-200 ng of purified DHFR
western blot (chemiluminescent): 0.5-1.0 μg/mL using 50-100 ng of purified recombinant DHFR, or 293T human embryonic kidney cell extract, or Jurkat human acute T Cell leukemia cell extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DHFR(1719)
mouse ... Dhfr(13361)
rat ... Dhfr(24312)

Specificity

DHFR and DHFR fusion proteins. The epitope resides within amino acids 171-185 of mouse DHFR.

Immunogen

amino acids 171-185 of mouse DHFR, conjugated to KLH via an N-terminal added cysteine residue.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gilad Barshad et al.
Scientific reports, 9(1), 9987-9987 (2019-07-12)
Mitochondrial complex I (CI) is the largest multi-subunit oxidative phosphorylation (OXPHOS) protein complex. Recent availability of a high-resolution human CI structure, and from two non-human mammals, enabled predicting the impact of mutations on interactions involving each of the 44 CI
Neda Habibi Arejan et al.
Frontiers in microbiology, 13, 1085918-1085918 (2023-01-31)
Mycobacterial cell elongation occurs at the cell poles; however, it is not clear how cell wall insertion is restricted to the pole or how it is organized. Wag31 is a pole-localized cytoplasmic protein that is essential for polar growth, but
Lauren A Howell et al.
Scientific reports, 9(1), 12418-12418 (2019-08-29)
The 26S proteasome conducts the majority of regulated protein catabolism in eukaryotes. At the heart of the proteasome is the barrel-shaped 20S core particle (CP), which contains two β-rings sandwiched between two α-rings. Whereas canonical CPs contain α-rings with seven

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service