Skip to Content
Merck
All Photos(1)

Key Documents

D044

Sigma-Aldrich

Amfonelic acid

powder

Synonym(s):

7-Benzyl-1-ethyl-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N2O3
CAS Number:
Molecular Weight:
308.33
MDL number:
UNSPSC Code:
12352200

form

powder

color

yellow

solubility

0.1 M NaOH: soluble
H2O: slightly soluble
ethanol: soluble

Biochem/physiol Actions

Blocks dopamine uptake

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R W Fuller et al.
Research communications in chemical pathology and pharmacology, 48(1), 17-25 (1985-04-01)
1-Methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) given in 4 daily s.c. injections to mice resulted in marked depletion of striatal dopamine, 3,4-dihydroxyphenylacetic acid (DOPAC) and homovanillic acid (HVA) 1 week after the last dose. Pretreatment of mice with alpha-methyltyrosine or Ro 4-1284 to deplete
C Pu et al.
Brain research, 649(1-2), 217-224 (1994-06-27)
Administration of methamphetamine (MA) induces degeneration of dopaminergic nerve terminals and astrogliosis, such as hypertrophy and an increase in apparent number, in the neostriatum. In this experiment adult rats were treated with MA (10 mg/kg, i.p.) 4 times in one
A V Juorio
British journal of pharmacology, 77(3), 511-515 (1982-11-01)
1 The concentrations of p- and m-tyramine, dopamine and homovanillic acid were measured in the mouse striatum following the subcutaneous administration of amfonelic acid, (+)-amphetamine or nomifensine.2 The administration of 2.5-25 mg/kg of amfonelic acid produced a reduction in p-tyramine
A G Ewing et al.
Science (New York, N.Y.), 221(4606), 169-171 (1983-07-08)
Microvoltammetric electrodes were used to monitor dopamine released in the caudate nucleus of the rat after electrical stimulation of the medial forebrain bundle. The time resolution of the technique is sufficient to determine in vivo concentration changes on a time

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service