Skip to Content
Merck

D0250

α-Dansyl-L-arginine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H25N5O4S · HCl
CAS Number:
Molecular Weight:
443.95
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
248-904-6
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (TLC)

Quality Segment

form

powder

color

yellow to green

storage temp.

−20°C

SMILES string

CN(C)c1cccc2c(cccc12)S(=O)(=O)NC(CCCNC(N)=N)C(O)=O

InChI

1S/C18H25N5O4S/c1-23(2)15-9-3-7-13-12(15)6-4-10-16(13)28(26,27)22-14(17(24)25)8-5-11-21-18(19)20/h3-4,6-7,9-10,14,22H,5,8,11H2,1-2H3,(H,24,25)(H4,19,20,21)

InChI key

HRBPBWKDJGGGCX-UHFFFAOYSA-N

Biochem/physiol Actions

α-Dansyl-L-arginine contains an N-terminal fluorophore (dansyl) making it useful as fluorescent marker/probe. α-Dansyl-L-arginine is used as a site specific marker for drug binding pockets on proteins. α-Dansyl-L-arginine, dansyl-l-asparagine, and dansyl-l-glutamate are specific for the site 1 drug binding site on albumin (HSA).


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library