CWDS-IF594
ColorWheel® Antibody-Ready iFluor 594 Dye
for use with ColorWheel® Antibody (Required, Sold Separately)
Synonym(s):
iFluor 594
About This Item
Recommended Products
product line
ColorWheel®
Quality Level
form
lyophilized
purified by
ion-exchange chromatography
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
technique(s)
flow cytometry: suitable
fluorescence
λex 588 nm; λem 604 nm
compatibility
for use with ColorWheel® Antibody (Required, Sold Separately)
greener alternative category
, Aligned
shipped in
ambient
storage temp.
2-8°C
target post-translational modification
unmodified
General description
iFluor 594 is optimized for labeling antibodies and proteins. It is a pH-independent dye that works over a wide range of pH (3 to 11). It displays an excitation max at 588 nm and an emission max at 604 nm. Its spectral properties are similar to Texas Red® and ALEXA FLUOR™ 594. However, compared to Texas Red®, it is easier to conjugate with R-Phycoerythrin (RPE) with higher conjugation yield and the resulting RPE-iFluor 594 tandem dye offers better FRET efficiency. It exhibits decent stability shows good reactivity and selectivity with amino groups.
Specificity
Application
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Physical form
CWDS-IF594-25UL - ColorWheel® Antibody-Ready iFluor 594 Dye Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.
CWD-FINISH-25UL - ColorWheel® Finishing Buffer Lyophilized from PBS. Contains no biocide or preservatives, such as azide.
Reconstitution
Storage and Stability
Legal Information
Disclaimer
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service