Skip to Content
Merck

Skip To

CRISPR18

Lenti CRISPR Universal Non-Target Control#1 Plasmid DNA (LV04 vector)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.51
UNSPSC Code:
41106609
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

packaging

vial of 50 μL

concentration

20 ng/μL in TE buffer; DNA (1μg of plasmid DNA)

application(s)

CRISPR
CRISPR

shipped in

dry ice

storage temp.

−20°C

General description

The Lenti CRISPR Non-Target Control#1 (LV04 vector) is a lentiviral plasmid vector, which includes a gRNA sequence that does not target known human, mouse and rat genes. The DNA format of this control construct permits direct transfection in knockout experiments or use in the viral packaging process (for lentiviral-based knockdown experiments). This vector is useful as a negative control in experiments using CRISPR lentiviral clones. The lenti CRISPR non-target control uses a dual-component system consisting of U6-driven guide RNA (non-targeting) and hPGK-driven Puromycin resistance cassette and Blue Fluorescence Protein for fluorophorescence. This control requires Cas9 be delivered by a separate vector.

Application

Functional Genomics/Screening/Target Validation

Other Notes

Ampicillin and puromycin antibiotic resistance genes provide selection in bacterial or mammalian cells, respectively. Self-inactivating replication incompetent viral particles can be produced in packaging cells (HEK293T) by co-transfection with compatible packaging plasmids (Product No. SHP002).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CRISPR18VCRISPR19VCRISPR20V
description

-

description

-

description

-

description

-

form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

−20°C

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

concentration

20 ng/μL in TE buffer; DNA (1μg of plasmid DNA)

concentration

(1x106 TU/ml (via p24 titering assay))

concentration

(1x106 TU/mL (via p24 titering assay))

concentration

(1x106 TU/mL (via p24 titering assay))

packaging

vial of 50 μL

packaging

vial of 200 μL

packaging

vial of 200 μL

packaging

vial of 200 μL

application(s)

CRISPR

application(s)

CRISPR

application(s)

CRISPR

application(s)

CRISPR


Storage Class

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters