Skip to Content
Merck
All Photos(1)

Key Documents

CLS480130

Corning® LSE single block, for Corning LSE single block digital dry baths

single block, 96 well PCR microplate, accommodates skirted or nonskirted plates, 1/cs

Synonym(s):

equipment replacement accessory

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€305.00

€305.00


Check Cart for Availability


Select a Size

Change View
1 EA
€305.00

About This Item

UNSPSC Code:
41102404
NACRES:
NB.15

€305.00


Check Cart for Availability

feature

Holds 1 x 96-well plates

packaging

case of 1
pack of 1

manufacturer/tradename

Corning 480130

technique(s)

PCR: suitable

suitability

suitable for (use with Corning LSE digital dry bath heater, single block)

Looking for similar products? Visit Product Comparison Guide

General description

Corning LSE single blocks are suitable for the following digital dry baths:

CLS6785SB, CLS6786SB, CLS6787SB

Legal Information

Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service