Key Documents
CLS480121
Corning® LSE single block, for Corning LSE single block digital dry baths
single block, 20 x 10 mm tubes, 1/cs
Synonym(s):
equipment replacement accessory
Select a Size
1 EA
€124.00
€124.00
Estimated to ship on29 May 2025
Select a Size
Change View
1 EA
€124.00
About This Item
UNSPSC Code:
41102404
NACRES:
NB.15
€124.00
Estimated to ship on29 May 2025
feature
Holds 20 x 10 mm tubes
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 480121
suitability
suitable for (use with Corning LSE digital dry bath heater, single block)
General description
Corning LSE single blocks are suitable for the following digital dry baths:
CLS6785SB, CLS6786SB, CLS6787SB
Legal Information
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)