CLS4519153
Corning® ProCulture® spinner flask accessories
Gas and Media Handling Fitting with Dual 1/8 in. Inlets for 8L Angled Sidearm Flasks
Synonym(s):
container accessory
Select a Size
€608.00
Select a Size
About This Item
€608.00
Recommended Products
manufacturer/tradename
Corning 4519-153
Related Categories
General description
Dual tubes extend to the bottom of the flask, and are held in place by Noryl nuts with integrated ferrules.
Combination tubes has two different length tubes, one extends to the bottom of the flask; the other is a shorter 6′ length. Both tubes are held in place by Noryl nuts with integrated ferrules.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)