CLS3772
Corning® microcarriers
non-treated
, weight 10 g
Synonym(s):
cell culture
Select a Size
€92.50
Select a Size
About This Item
€92.50
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Corning 3772
weight
10 g
General description
Features:
- USP Class VI polystyrene material provides a consistent platform
- Level (SAL) of 10-6
- Certified nonpyrogenic
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)