CLS3721
Corning® 96 Well Half-Area Microplate
flat bottom clear, black polystyrene, Tissue Culture (TC)-treated surface, bag of 5, sterile, lid
All Photos(1)
About This Item
Recommended Products
material
black polystyrene
clear bottom
flat bottom clear
sterility
sterile
feature
lid
skirt
packaging
bag of 5
manufacturer/tradename
Corning 3721
size
96 wells
well D
10.7 mm
well volume
190 μL
well working volume
25-125 μL
binding type
Tissue Culture (TC)-treated surface
General description
Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.
Legal Information
Corning is a registered trademark of Corning, Inc.