Skip to Content
Merck
All Photos(1)

Key Documents

CLS358339

Corning® Falcon® Cell Strainer For Use With 50ml Conical Tubes

size 40 μm, blue Nylon, non-sterile

Synonym(s):

culture tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104932
NACRES:
NB.13
Pricing and availability is not currently available.

material

blue Nylon

sterility

non-sterile

manufacturer/tradename

Corning 358339

size

40 μm

application(s)

filtration

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service