Skip to Content
Merck
All Photos(1)

Key Documents

CLS3576

Corning® 384 well microplate, low flange

high binding surface, white polystyrene, flat bottom, non-sterile, lid: no, pack of 50

Synonym(s):

384 multiwell plates, 384 well immunoassay plates, 384 well microplates, 384 well microtiter plates, 384 well plates, immunoassay plates

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
€648.00

€648.00


Check Cart for Availability


Select a Size

Change View
50 EA
€648.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
flat bottom
polystyrene
square white wells
white bottom
white polystyrene
size:
384 wells

€648.00


Check Cart for Availability

material

flat bottom
polystyrene
square white wells
white bottom
white polystyrene

sterility

non-sterile

feature

lid: no
skirt
plate format: 384 well standard

packaging

pack of 50

manufacturer/tradename

Corning 3576

maximum volume

112 μL

size

384 wells

working volume

80 μL

color

white

suitability

suitable for (cell cuture)

binding type

high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Designed to reduce well-to-well crosstalk during luminescent assays
  • Recommended working volume of up to 20 to 80μL
  • Conforms to standard microplate footprint and dimensions
  • High binding surface binds medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions
  • Binding capacity is approximately 400 to 500ng IgG/cm2
  • Ideal for immunoassays

Note: Suggested replacement for CLS3703

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service