Skip to Content
Merck
All Photos(4)

Key Documents

CLS356258

Corning® Matrigel® Matrix -3D Microplate

well size 384 wells, clear white wells, lid, Phenol Red-Free

Synonym(s):

Cell Culture Plate, plastic coated cell culture plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.13
material:
clear white wells
size:
384 well
sterility:
sterile
binding type:
Tissue Culture (TC)-treated surface
feature:
lid
Pricing and availability is not currently available.

material

clear white wells

sterility

sterile

feature

lid

packaging

pkg of 1 ea (Individually wrapped)

manufacturer/tradename

Corning 356258

size

384 well

well size

384 wells

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

Application

Cell culture

Legal Information

Corning is a registered trademark of Corning, Inc.
Matrigel is a registered trademark of Corning, Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service