Key Documents
CLS3514
Corning® FiltrEX plates
polystyrene non-sterile, 100 ea
Synonym(s):
96 multiwell plate
About This Item
UNSPSC Code:
41121800
NACRES:
NB.24
Pricing and availability is not currently available.
Product Name
Fluid Guard for Corning® 96 well FiltrEX plates, polystyrene, non-sterile
material
polystyrene
sterility
non-sterile
manufacturer/tradename
Corning 3514
General description
The fluid guard is manufactured from thermoformed polystyrene, non-sterile and easily snaps to the base of the filter plate. The unique design enables the guard to be applied manually or robotically to the plate. The guard serves to protect laboratory furniture and equipment from hazardous or radioactive materials.
Legal Information
Corning is a registered trademark of Corning, Inc.
FiltrEX is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)